ContactUsLinkComponent
NALGENE* Drop-Dispenser Bottles, Low-Density Polyethylene
Nalgene® Drop-Dispenser Bottles, Low-Density Polyethylene, Thermo Scientific
# de catalogue 16354-421
NALGENE* Drop-Dispenser Bottles, Low-Density Polyethylene
Nalgene® Drop-Dispenser Bottles, Low-Density Polyethylene, Thermo Scientific
# de catalogue 16354-421
Spécifications
  • Capacity:
    30 ml (1 oz.)
  • Cat. no.:
    16354-421
  • Supplier No.:
    2411-0030
Spécifications
A propos de cet article
Bottles dispense one drop at a time.
Captive spout cap won't release or leak when bottle is squeezed.
Delivery: Supplied with polypropylene dropping closure and spout plug.
Customers Who Bought This Also Bought