WHEATON® Dropper Tip Caps, DWK Life Sciences
# de catalogue 16300-114
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Cap Size:15-415
- Color:Red
- Cat. no.:16300-114
- Supplier No.:242512
Spécifications
A propos de cet article
Dropper tip caps are made of radiation grade polypropylene in a variety of colors
Packaged in cases of 100, 1000, or bulk.
Customers Who Bought This Also Bought