Nalgene® Unitary™ Vented Safety Wash Bottles, Low-Density Polyethylene, Thermo Scientific
# de catalogue 16210-925
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:500 ml (16.9 oz.)
- Description:Sodium Hypochlorite (Bleach)
- Cat. no.:16210-925
- Cap Color:White
- Supplier No.:2436-0506
Spécifications
A propos de cet article
LDPE bottles feature a unique vent system to keep hazardous volatile liquids in the bottle and off the lab bench.
Sodium hypochlorite (bleach) bottles are supplied with vented, white HDPE 38 mm closure; all other bottles supplied with vented polypropylene 38 mm colored closure. To prevent cross-contamination, each bottle has a color bar below the chemical name that corresponds to the colored closure.
Unitary design prevents contamination when filling the bottle. A strong, steady stream of liquid is dispensed when the bottle is squeezed.
Customers Who Bought This Also Bought