Order Entry
ContactUsLinkComponent
PYREX* Media Bottle, Graduated
PYREX® Media/Storage Bottles, Narrow Mouth, Round, with Screw Cap, Corning
# de catalogue 16157-136
Fournisseur:  Corning
Product available on GSA Advantage®
PYREX* Media Bottle, Graduated
PYREX® Media/Storage Bottles, Narrow Mouth, Round, with Screw Cap, Corning
# de catalogue 16157-136
Fournisseur:  Corning
Product available on GSA Advantage®

Spécifications

  • Cap Size:
    GL45
  • Capacity:
    250 ml (8.5 oz.)
  • Graduation Interval:
    50 mL
  • Graduation range:
    50–200 mL
  • Supplier No.:
    1395-250

Spécifications

A propos de cet article

Heavy duty bottles, made of PYREX® borosilicate glass, with orange PP linerless, one-piece autoclavable plug seal screw with a drip-free pouring ring.

  • Permanent white enamel graduations with marking spots
  • Glass bead indicates full capacity line
  • Very good temperature resistance, very good chemical resistance

Graduated, are used for storage as well as mixing and sampling.

Further colour-coded PP caps, PBT caps, ETFE pouring rings (for dry heat sterilization (180 °C)), and vented membrane caps are available.

Caution: Do not tighten cap immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Bottles larger than 2 L should not be used with bottle top filter units or in other applications involving vacuum pressure as breakage may occur. A vented cap is not recommended for use on the 5 and 10 L bottles for venting during autoclaving.
Customers Who Bought This Also Bought