Nalgene® Carboys with Handles, Wide Mouth, Low-Density Polyethylene, Thermo Scientific
# de catalogue 16133-025
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:10 L (2.6 gal.)
- Supplier No.:2234-0020
Spécifications
A propos de cet article
For corrosive liquids and solids.
- Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Wide mouth allows easy access for cleaning. Convenient shoulder handles. Graduated to contain in 1 gal. and 5 L increments. Cap Size: 100-415.
Customers Who Bought This Also Bought