Nalgene® Heavy-Duty Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
# de catalogue 16129-630
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:53B
- Capacity:2 L (67.6 oz.)
- Cat. no.:16129-630
- Supplier No.:2125-2000
Spécifications
A propos de cet article
Rigid, thick-walled bottles are designed for use as extra-rugged storage, reservoirs, or shipping containers for liquids or dry chemicals.
- Leakproof
Ordering information: For replacement closures, see 16219-122 series.
Delivery: Supplied with white polypropylene heavy-duty closures.
Customers Who Bought This Also Bought