VWR® Wide Mouth Bottles, Round, HDPE,
Catalog # 16125-843
Supplier: VWR International
Small Business Enterprise
Specifications
- Cap size:53-400
- Capacity:500 ml (16.9 oz.)
- Cat. no.:16125-843
Specifications
About this item
Lightweight bottles are an excellent value for storage of both liquid and powdered process or environmental samples.
- Lightweight
- HDPE body construction
- Polypropylene caps
- Wide-mouthed design
White polypropylene caps with polyethylene film/foam liners eliminate leaks.
Customers Who Bought This Also Bought