VWR® Wide Mouth Bottles, Round, HDPE,
# de catalogue 16125-785
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Cap Size:38-400
- Capacity:120 ml (4 oz.)
- Cat. no.:16125-785
Spécifications
A propos de cet article
Lightweight bottles are an excellent value for storage of both liquid and powdered process or environmental samples.
- Lightweight
- HDPE body construction
- Polypropylene caps
- Wide-mouthed design
White polypropylene caps with polyethylene film/foam liners eliminate leaks.
Customers Who Bought This Also Bought