ContactUsLinkComponent
VWR* Round Bottles, High-Density Polyethylene, Wide Mouth
VWR® Wide Mouth Bottles, Round, HDPE,
# de catalogue 16125-752
Fournisseur:  VWR International
Small Business Enterprise
VWR* Round Bottles, High-Density Polyethylene, Wide Mouth
VWR® Wide Mouth Bottles, Round, HDPE,
# de catalogue 16125-752
Fournisseur:  VWR International
Small Business Enterprise
Spécifications
  • Cap Size:
    33-400
  • Capacity:
    60 ml (2 oz.)
  • Cat. no.:
    16125-752
Spécifications
A propos de cet article
Lightweight bottles are an excellent value for storage of both liquid and powdered process or environmental samples.
  • Lightweight
  • HDPE body construction
  • Polypropylene caps
  • Wide-mouthed design
White polypropylene caps with polyethylene film/foam liners eliminate leaks.
Customers Who Bought This Also Bought