Nalgene® Unitary™ Color-Coded Wash Bottles, Low-Density Polyethylene, Thermo Scientific
# de catalogue 16125-170
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:500 ml (16.9 oz.)
- Cat. no.:16125-170
- Supplier No.:2423-0500
Spécifications
A propos de cet article
Covenient, pre-packaged Unitary™ wash bottles come with four polypropylene color closures (one each of white, red, yellow, and blue) for easy identification.
Ordering information: A sample of PolyPaper Right-To-Know Custom Labeling System with Mylar® overlay is included in each pack.
Customers Who Bought This Also Bought