Nalgene® Square Laboratory Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
# de catalogue 16120-753
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:38-430
- Capacity:1 L (33.8 oz.)
- Cat. no.:16120-753
- Supplier No.:2016-1000
Spécifications
A propos de cet article
Square laboratory bottles provide excellent chemical resistance.
- Leakproof
- Repeatedly autoclavable
Note: Before autoclaving, set closure on top of the bottle without engaging the threads.
The translucent design is ideal for long-term storage of inorganic buffers, alcohols, acids, and bases. All sizes have permanent graduations in convenient increments. Bottles are designed for use within a temperature range of 0 to 135 °C (32 to 275 °F).
Ordering information: Supplied with polypropylene screw closures assembled on bottles. For replacement caps for 16120-741 through -753, see 16219-122 series.
Customers Who Bought This Also Bought