Order Entry
ContactUsLinkComponent
NALGENE* Carboys with Spigot and Handles, Polypropylene
Nalgene® Carboys with Spigot and Handles, Polypropylene, Thermo Scientific
# de catalogue 16101-448
NALGENE* Carboys with Spigot and Handles, Polypropylene
Nalgene® Carboys with Spigot and Handles, Polypropylene, Thermo Scientific
# de catalogue 16101-448

Spécifications

  • Capacity:
    20 L (5.3 gal.)
  • Cat. no.:
    16101-448
  • Supplier No.:
    2319-0050

Spécifications

A propos de cet article

Use for storing and dispensing solutions and media.

  • Provide excellent chemical resistance

Quick Action spigot boss is molded in one piece directly to the body of the carboy for strength and leak-resistance. Cap size: 83B. Also takes No. 13¹/₂ stopper.

For replacement caps, see cap 16225-234 in 16219-122 series.

Note: Autoclave using a properly vented closure. Disassemble spigot before autoclaving.

Ideal for sterile water storage, carboys can be autoclaved before refilling to stop recurring algae or bacterial growth. Graduated to deliver in 1 gal. and 5 L increments.

Ordering information: Supplied with carboy, polypropylene spigot, and screw cap.
Customers Who Bought This Also Bought