Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
Catalogue # 16093-011
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:38-430
- Capacity:4 L (1 gal.)
- Cat. no.:16093-011
- Supplier No.:2221-0010
Spécifications
À propos de cet article
Autoclavable, leakproof jugs offer excellent chemical resistance.
- Leakproof
- Autoclavable
Note: Before autoclaving, set cap on top of jug without engaging the threads.
A large carrying handle allows for the use of rubber gloves for safe handling of corrosive liquids, as well as an offset pour spout for easy, accurate pouring.
Ordering information: Jugs are supplied with polypropylene screw closures. For replacement caps, see 16219-122 series
Frequently Bought Together