VWR® Narrow Mouth Sample Bottles, HDPE
# de catalogue 16058-848
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Cap Size:24-410
- Capacity:500 ml (16.9 oz.)
- Cat. no.:16058-848
Spécifications
A propos de cet article
Lightweight, economy bottles offer an outstanding value for high volume sampling requirements.
- Lightweight
- Economical
- Narrow-mouthed design
- HDPE construction
- Straight sides
Bottles have a straight-side commercial design. Tough polypropylene caps with polyethylene foam liners ensure tight seals.
Customers Who Bought This Also Bought