Nalgene® Large Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Catalog # 16058-189
Supplier: Thermo Fisher Scientific
Specifications
- Cap size:38-430
- Capacity:4 L (1 gal.)
- Cat. no.:16058-189
- Supplier No.:2202-0010
Specifications
About this item
Large bottles are designed for collecting, storing, or shipping anything from distilled water to corrosive chemicals and standards.
- With polypropylene closures
- Leakproof
A built-in loop on the shoulder (except on the 2 L size) is convenient for attaching an identification tag.
Ordering information: For replacement closures, see 16219-122 series.
Customers Who Bought This Also Bought