Order Entry
ContactUsLinkComponent
NALGENE* Large Bottles, Low-Density Polyethylene, Narrow Mouth
Nalgene® Large Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
# de catalogue 16058-167
NALGENE* Large Bottles, Low-Density Polyethylene, Narrow Mouth
Nalgene® Large Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
# de catalogue 16058-167

Spécifications

  • Cap Size:
    38-430
  • Capacity:
    2 L (67.6 oz.)
  • Cat. no.:
    16058-167
  • Supplier No.:
    2202-0005

Spécifications

A propos de cet article

Large bottles are designed for collecting, storing, or shipping anything from distilled water to corrosive chemicals and standards.

  • With polypropylene closures
  • Leakproof

A built-in loop on the shoulder (except on the 2 L size) is convenient for attaching an identification tag.

Ordering information: For replacement closures, see 16219-122 series.
Customers Who Bought This Also Bought