ContactUsLinkComponent
VWR* Hemacytometer Cover Glass
VWR® Hemacytometer Cover Glasses
# de catalogue 16007-000
Fournisseur:  VWR International
Product available on GSA Advantage®
VWR* Hemacytometer Cover Glass
VWR® Hemacytometer Cover Glasses
# de catalogue 16007-000
Fournisseur:  VWR International
Product available on GSA Advantage®
Spécifications
  • Description:
    Cover glass hemacytometer, rectangular
  • L×W:
    26×20 mm
  • Thickness:
    0.4 mm
Spécifications
A propos de cet article
Highly polished, hard-ground glass hemacytometer cover glass features rectangular edges with clipped corners
Highly polished, hard-ground glass hemacytometer cover glass features rectangular edges with clipped corners
The cover glass is transparent.
Customers Who Bought This Also Bought