Order Entry
Canada
ContactUsLinkComponent
VWR® Disposable Aluminum Crinkle Dishes with Tabs
Catalogue # 16005-126
Fournisseur:  VWR International
undefined
VWR® Disposable Aluminum Crinkle Dishes with Tabs
Catalogue # 16005-126
Fournisseur:  VWR International

Spécifications

  • Capacity:
    60 ml
  • Cat. no.:
    16005-126
  • I.D.×D:
    5.7×1.6 cm (2¼×⅝")
  • Weight:
    1.3 g

Spécifications

À propos de cet article

These dishes are ideal for general-purpose weighing, dispensing, and storing applications.

  • Dishes feature tabs for easy handling
  • Crimped sides for rigidity
  • Sturdy construction

Dishes also function as evaporating dishes, parts containers, or dust covers. Sturdy, with a flat bottom and slight taper for easy stacking.

Frequently Bought Together