ContactUsLinkComponent
VWR® Premium VistaVision™ Plain and Frosted Microscope Slides
Catalogue # 16004-430
Fournisseur:  VWR International
Product available on GSA Advantage®
undefined
VWR® Premium VistaVision™ Plain and Frosted Microscope Slides
Catalogue # 16004-430
Fournisseur:  VWR International
Product available on GSA Advantage®
Spécifications
  • Description:
    Slides
  • L×W:
    3×1"
  • Thickness:
    1.0 mm
  • Cat. no.:
    16004-430
Spécifications
À propos de cet article
Microscope slides are manufactured of soda-lime glass in accordance with ISO 8037/1 standards.
  • Manufactured in Germany using high-quality, white soda lime glass
  • Pre-cleaned and ready for use
  • Autoclavable
  • Plain and frosted formats available
A thorough automatic cleaning process ensures that slides are ready to use without further treatment. Frosted slides feature a fine 20 mm frosted marking area.
Frequently Bought Together