VWR® VistaVision™ HistoBond® Premium Adhesion Slides
Catalogue # 16004-406
Fournisseur: VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Spécifications
- Color:White
- L×W:75×25" mm (3×1")
- Thickness:1 mm
- Cat. no.:16004-406
Spécifications
À propos de cet article
VWR® VistaVision™ HistoBond® Adhesive Slides are ideal for in-situ hybridization.
- Manufactured in Germany using high-quality, white soda lime glass
- Pre-cleaned and ready for use
- Autoclavable
- Color-coded ends facilitate sorting and identification
- Uniform size and thickness
The slide surface is loaded with a permanent positive charge that firmly anchors all types of sections. This treatment prevents the loss of tissue sections or cells to frequent washing, and eliminates the disturbing background effect caused by traditional adhesives.
Frequently Bought Together