Order Entry
ContactUsLinkComponent
VWR*VistaVision* Cover Glasses, No. 1 1/2
VWR VistaVision™ Cover Glasses, No. 11/2
Catalog # 16004-340
Supplier:  VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
VWR*VistaVision* Cover Glasses, No. 1 1/2
VWR VistaVision™ Cover Glasses, No. 11/2
Catalog # 16004-340
Supplier:  VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise

Specifications

  • Description:
    Cover glasses, rectangular
  • L×W:
    40×24 mm
  • Thickness:
    0.16 - 0.19 mm
  • Cat. no.:
    16004-340

Specifications

About this item

Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.

  • Pure white borosilicate glass fabrication
  • Excellent chemical resistance and flatness

Under ultraviolet light, they emit no disturbing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.

Refractive index: 1.525 ±0.0015 ne

Ordering information: Square and rectangular cover glasses are supplied 1 oz. per two-part box; rectangular cover glasses are supplied 1 oz. per hinged boxes.
Customers Who Bought This Also Bought