Order Entry
ContactUsLinkComponent
VWR*VistaVision* Cover Glasses, No. 1
VWR VistaVision™ Cover Glasses, No. 1
Catalog # 16004-328
Supplier:  VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
VWR*VistaVision* Cover Glasses, No. 1
VWR VistaVision™ Cover Glasses, No. 1
Catalog # 16004-328
Supplier:  VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise

Specifications

  • Numbering:
    #1
  • Description:
    Cover glasses, square
  • L×W:
    25×25 mm
  • Thickness:
    0.13 - 0.16 mm
  • Cat. no.:
    16004-328

Specifications

About this item

Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.

  • Pure white borosilicate glass fabrication
  • Excellent chemical resistance and flatness

Under ultraviolet light, they emit no disturbing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.

Refractive index: 1.525 ±0.0015 ne

Ordering information: Circular and square cover glasses are supplied 100 per two-part box; rectangular cover glasses are supplied 100 per hinged boxes.
Customers Who Bought This Also Bought