ContactUsLinkComponent
U-TEK* Reusable Refrigerant Gel Pack
U-tek® Gel Packs, Phase Change Material Gel Packs, Sonoco ThermoSafe
Catalog # 15715-137
Supplier:  Sonoco Thermosafe
undefined
U-tek® Gel Packs, Phase Change Material Gel Packs, Sonoco ThermoSafe
Catalog # 15715-137
Supplier:  Sonoco Thermosafe
Specifications
  • L×W×D:
    15 x 10 x 2 cm (6 x 4 x 3/4")
  • Weight:
    227 g (8 oz.)
  • Supplier No.:
    432
Specifications
About this item
U-TEK® phase change materials (PCM) are formulated refrigerant gel packs for products that require frozen temperatures, but cannot tolerate dry ice or CO2.
  • For products that cannot tolerate dry ice
  • Extended protection
  • Radiant heat barrier
An alternative to water-based ice packs, these coolant packs offer superior protection for extended periods. Laminated, metallized film exterior provides radiant heat barrier and resists punctures. Ideal for use with ThermoSafe foam mailers.
Frequently Bought Together