Order Entry
Canada
ContactUsLinkComponent
Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Catalogue # 15713-610
Fournisseur:  Sonoco Thermosafe
undefined
Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Catalogue # 15713-610
Fournisseur:  Sonoco Thermosafe
Supplier Number:  440

Spécifications

  • Description:
    Holds vials, tubes, Small bottles, tissue flasks with growth area of 75 cm²
  • Cat. no.:
    15713-610
  • Interior Dimensions:
    14.9L×11.1W×7.6H cm (5 7/8×4 3/8×3")
  • Supplier No.:
    440

Spécifications

À propos de cet article

Lightweight, economical EPS shippers provide maximum breakage protection for known non-hazardous materials during specimen transport.

  • For VacuTainer®, Microtainer®, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Lightweight polystyrene foam saves postage
  • Reusable and recyclable

A unique array of specimen shippers fills the need for protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.

Frequently Bought Together