ContactUsLinkComponent
Double Neubauer Counting Chamber and Cover
Double Neubauer Counting Chamber Set, Hausser Scientific
# de catalogue 15170-173
Fournisseur:  Hausser
Double Neubauer Counting Chamber Set, Hausser Scientific
# de catalogue 15170-173
Fournisseur:  Hausser
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chamber and Cover Glass (2) Only
  • Cat. no.:
    15170-173
  • Chamber Depth:
    0.1 mm
  • Supplier No.:
    3100
Spécifications
A propos de cet article
Optically ground and polished milled glass chambers are diamond etched with Double improved Neubauer ruling
A “V” slash on each plateau facilitates charging and reduces the possibility of overflow. Metal plating on the plateaus provides sharp bright line contrast for consistent cell counts. Triple dividing lines clearly show cells within the counting area. Ruled surface is 0.1mm below the cover glass. Fluid capacity is 0.1mm3/mm2 and 0.00025mm3/each grid square.
Ordering information: Set includes counting chamber, two cover glasses, one red cell pipet, one white blood cell pipet, pipettor filler, and plastic carrying case. For additional cover glasses compatible with the Double Neubauer Counting Chamber Set, see the Hemacytometer Cover Glasses, Hausser Scientific series (23649-035).
Customers Who Bought This Also Bought