VWR® Rubber Bulbs
# de catalogue 15001-362
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Capacity:1 ml
- Color:Black
- Cat. no.:15001-362
Spécifications
A propos de cet article
Made of durable natural rubber, bulbs are suitable for medicine droppers and small dropping pipettes.
- Durable
- Natural rubber construction
- Heavy-walled design
- Cylindrical
Neck is grooved to take flat flange of pipettes. Bulbs fit pipettes 6 to 8 mm ext. Ø.
Customers Who Bought This Also Bought