Order Entry
ContactUsLinkComponent
Micro-Hematocrit Capillary Tubes
Micro-Hematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Catalog # 14705-003
Micro-Hematocrit Capillary Tubes
Micro-Hematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Catalog # 14705-003

Specifications

  • Volume:
    70 µl
  • Color:
    Red
  • Description:
    Microhematocrit Capillary tube
  • Length:
    75 mm
  • Cat. no.:
    14705-003
  • Inner Diameter:
    1.1 mm
  • Supplier No.:
    41B2501

Specifications

About this item

These Micro-Hematocrit Capillary Tubes are manufactured from soda lime glass, packed in plastic vials with reclosable snap-lock caps to maintain cleanliness.

Ends may be sealed by flame, sealing clay or patented caps. Heparinized capillary tubes are treated to prevent blood clotting.

Customers Who Bought This Also Bought