Samco™ Disposable Transfer Pipettes, Fine Tip, Thermo Scientific
Catalogue # 14670-330
Fournisseur: Thermo Fisher Scientific
Spécifications
- Sterility:Non sterile
- Capacity:5.8 ml
- Description:Standard Fine Tip
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Packed:Bulk
- Cat. no.:14670-330
- Overall Length:14.7 cm (5²⁵⁄₃₂")
- Stem Diameter:4.6 mm
- Bulb Draw:3.3 ml
- Supplier No.:232-11
Spécifications
À propos de cet article
These fine-tip polyethylene transfer pipettes are excellent for the precise dispensing of small drops (45 to 50 per ml) into microbiological Int.Ø.
Strips, ELISA plates, and microcentrifuge and precipitation tubes. Ideal for removing cell layers, filling electrophoresis wells, spotting chromatography plates, and other similar applications.
Frequently Bought Together