Order Entry
ContactUsLinkComponent
MicroReach* Guard Barrier Pipet Tips, Sterile
MµltiGuard™ Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 14234-098
Fournisseur:  Sorenson Bioscience
MµltiGuard™ Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 14234-098
Fournisseur:  Sorenson Bioscience

Spécifications

  • Length:
    4.5 cm (1.77")
  • Volume:
    0.1 - 10 µl
  • Tip Style:
    MicroReach™ Guard Tips, Graduated, Low Binding
  • Supplier No.:
    38000T

Spécifications

A propos de cet article

Ideal for PCR, radioisotope labeling, and pipetting procedures that require an aerosol-free method for handling samples.

  • Tips fit most popular research-grade pipettors

Low-binding tips feature reduced surface tension which helps minimize the bonding of DNA and protein molecules to the tip material.

Ordering information: Supplied in polypropylene CellRacks. MµltiGuard™ tips 14234-104 and 14234-106 can accommodate a volume of 30 µl.
Packaging: Packs contain 10 racks with 96 tips/rack, except 14234-120 and 14234-122, which have 100 tips/rack.
Customers Who Bought This Also Bought