ContactUsLinkComponent
Graduated Yellow Tips
MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Catalog # 14229-832
MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Catalog # 14229-832
Specifications
  • Volume:
    1 - 200 µl
  • Tip Color:
    Yellow
  • Packed:
    Bulk
  • Cat. no.:
    14229-832
  • Tip Style:
    Graduated
  • Supplier No.:
    15730
Specifications
About this item
Pipette tips feature a unique beveled end to minimize fluid retention.
  • Fully autoclavable
  • Tips fit most popular research-grade pipettors
They ensure superior sample delivery and recovery in critical micro-volume ranges. Reinforced base design and standard 96-well pattern of cell racks ensures rapid mounting of small tips on all multichannel pipettors.
Certifications: The 1 to 200 µl racked tips are certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Ordering information: Available in bulk packaging, autoclavable polypropylene racks, hinged racks, QuickRack reloads for reloading standard racks, and QuickShot reloads for reloading hinged racks.
Frequently Bought Together