MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Catalog # 14229-824
Supplier: Sorenson Bioscience
Specifications
- Volume:1 - 200 µl
- Tip Color:Natural
- Packed:Rack
- Cat. no.:14229-824
- Sterile:+
- Tip Style:Graduated
- Supplier No.:15670
Specifications
About this item
Pipette tips feature a unique beveled end to minimize fluid retention.
- Fully autoclavable
- Tips fit most popular research-grade pipettors
They ensure superior sample delivery and recovery in critical micro-volume ranges. Reinforced base design and standard 96-well pattern of cell racks ensures rapid mounting of small tips on all multichannel pipettors.
Certifications: The 1 to 200 µl racked tips are certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Ordering information: Available in bulk packaging, autoclavable polypropylene racks, hinged racks, QuickRack reloads for reloading standard racks, and QuickShot reloads for reloading hinged racks.
Frequently Bought Together