TWD Scientific Weighing Funnels
Catalog # 14221-828
Supplier: TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Capacity:2 ml
- Material:Borosilicate glass
- Size:Small
- Cat. no.:14221-828
- Supplier No.:GWF-BS1-S
- Fits:5, 10, 25, 50 ml volumetric flasks
Specifications
About this item
The TWD Scientific (formerly Tradewinds Direct) family of weighing funnels (weighing boats) all feature a flat bottom for sitting on balances and a pouring neck that fits to volumetric flasks, micro-tubes, or any narrow neck apparatus for spill-free transfer.
- Available in antistatic polypropylene, disposable polypropylene, and borosilicate glass
- Residual samples can be rinsed to guarantee 100% transfer
- Five different sizes of funnels fit to various flask sizes
- X-Small funnels fit 96-well mini-tubes and deep well plates
Suitable for safe handling of hazardous materials.
Customers Who Bought This Also Bought