TWD Scientific Weighing Funnels
# de catalogue 14221-812
Fournisseur: TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Capacity:2 ml solid/1 ml liquid
- Material:PP; anti-static
- Size:X-small
- Fits:96-well mini tubes, deep well plates
- Supplier No.:ASWFXS
Spécifications
A propos de cet article
The TWD Scientific (formerly Tradewinds Direct) family of weighing funnels (weighing boats) all feature a flat bottom for sitting on balances and a pouring neck that fits to volumetric flasks, micro-tubes, or any narrow neck apparatus for spill-free transfer.
- Available in antistatic polypropylene, disposable polypropylene, and borosilicate glass
- Residual samples can be rinsed to guarantee 100% transfer
- Five different sizes of funnels fit to various flask sizes
- X-Small funnels fit 96-well mini-tubes and deep well plates
Suitable for safe handling of hazardous materials.
Customers Who Bought This Also Bought