ContactUsLinkComponent
VistaLab* Ovation* pipette tips
Disposable Tips for MLA and Ovation Pipettors, VistaLab
# de catalogue 14220-735
Fournisseur:  VistaLab
Disposable Tips for MLA and Ovation Pipettors, VistaLab
# de catalogue 14220-735
Fournisseur:  VistaLab
Spécifications
  • Volume:
    25 µl
  • Packed:
    VistaBulk™
  • Tip Style:
    Micro
  • Supplier No.:
    4058-1000
Spécifications
A propos de cet article
Disposable polypropylene tips for use with MLA or ovation pipettors.
  • Non-pyrogenic
  • Non-sterile or sterile, RNase/DNase-free
  • Filtered or non-filtered
  • Low retention available
  • Trace metal certified available
VistaLab tips are available in a full range of volumes from 2 μl to 10 ml for Macro Volume pipettors.
These disposable tips are an economical choice and are available in a wide range of sizes.
Customers Who Bought This Also Bought