VWR® Autoclavable Bags, 1.5 mil
# de catalogue 14220-002
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- W×L:305×610 mm (12×24")
- Color:Red, printed
Spécifications
A propos de cet article
Autoclave bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
- Bags are available clear or red with proper biohazard warnings, or clear without label
- Rated for use at 135 °C (275 °F)
High-strength, highly temperature-resistant polypropylene construction allows perfect adaptability to standard laboratory disposal routines, including steam autoclaving.
Ordering information: Twist ties included.
Customers Who Bought This Also Bought