Pipette Tips, Sorenson BioScience
# de catalogue 14006-914
Fournisseur: Sorenson Bioscience
Spécifications
- Volume:0.1 - 10 µl
- Tip Color:Natural
- Sterile:+
- Tip Style:MicroReach
- Supplier No.:23570
Spécifications
A propos de cet article
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Customers Who Bought This Also Bought