ContactUsLinkComponent
MBP Gel Loading Pipette Tips, 200 µl, Thermo Scientific
Catalogue # 14004-776
undefined
MBP Gel Loading Pipette Tips, 200 µl, Thermo Scientific
Catalogue # 14004-776
Supplier Number:  010-R204
Spécifications
  • Volume:
    1 - 200 µl
  • Packed:
    Rack with Lift-off Lid
  • Cat. no.:
    14004-776
  • Sterile:
    -
  • Supplier No.:
    010-R204
Spécifications
À propos de cet article
Useful for a wide variety of sample loading applications such as protein separation, nucleic acid sequencing, and other gel loading applications.
  • Pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes
  • All materials used in the manufacture of our products are free of trace metals
  • All sterile products are sterilized in accordance with AAMI guidelines by gamma irradiation
For use with but not limited to: Pipetman®, Costar, Eppendorf®, Finnpipette®, Sealpette, Labpette™, Nichiryo, Oxford-Benchmate®, Rainin® EDP2, Socorex®, Titertek® Digital, and GeneMate.
Frequently Bought Together