VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Catalog # 13916-820
Supplier: VWR International
Specifications
- Graduation Range:250 - 5000 ml
- Capacity:5 L (169 oz.)
Specifications
About this item
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
- Molded of autoclavable, translucent polypropylene
- Low form for better stability
- All sizes graduated in milliliters and liters
Frequently Bought Together