VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
# de catalogue 13916-762
Fournisseur: VWR International
Spécifications
- Capacity:2 L (68 oz.)
- Graduation range:50 - 2000 ml
- Cat. no.:13916-762
Spécifications
A propos de cet article
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
- Molded of autoclavable, translucent polypropylene
- Low form for better stability
- All sizes graduated in milliliters and liters
Customers Who Bought This Also Bought