VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Catalog # 13916-707
Supplier: VWR International
Specifications
- Graduation range:25 - 500 ml
- Capacity:500 ml (17 oz.)
- Cat. no.:13916-707
Specifications
About this item
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
- Molded of autoclavable, translucent polypropylene
- Low form for better stability
- All sizes graduated in milliliters and liters
Customers Who Bought This Also Bought