VWR®, Griffin Beakers, Polypropylene
Catalog # 13890-148
Supplier: VWR International
Specifications
- Graduation range:100 - 1000 ml
- Capacity:1 L (34 oz.)
- Cat. no.:13890-148
Specifications
About this item
Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.
- Stackable
- Sturdy construction withstands repeated autoclaving
- Break-resistant, non-toxic, non-contaminating, and extremely chemical-resistant
- Feature no-drip spout for clean pouring
- Tapered walls for secure grip
- Oval marking spot
Ideal beaker for general laboratory procedures.
Customers Who Bought This Also Bought