ContactUsLinkComponent
VWR* Griffin Beakers, Polypropylene
VWR®, Griffin Beakers, Polypropylene
Catalog # 13890-046
Supplier:  VWR International
VWR* Griffin Beakers, Polypropylene
VWR®, Griffin Beakers, Polypropylene
Catalog # 13890-046
Supplier:  VWR International
Specifications
  • Graduation range:
    10 - 100 ml
  • Capacity:
    100 ml (3.4 oz.)
  • Cat. no.:
    13890-046
Specifications
About this item
Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.
  • Stackable
  • Sturdy construction withstands repeated autoclaving
  • Break-resistant, non-toxic, non-contaminating, and extremely chemical-resistant
  • Feature no-drip spout for clean pouring
  • Tapered walls for secure grip
  • Oval marking spot
Ideal beaker for general laboratory procedures.
Customers Who Bought This Also Bought