VWR®, Griffin Beakers, Polypropylene
# de catalogue 13890-024
Fournisseur: VWR International
Spécifications
- Capacity:50 ml (1.7 oz.)
- Graduation range:10 - 50 ml
- Cat. no.:13890-024
Spécifications
A propos de cet article
Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.
- Stackable
- Sturdy construction withstands repeated autoclaving
- Break-resistant, non-toxic, non-contaminating, and extremely chemical-resistant
- Feature no-drip spout for clean pouring
- Tapered walls for secure grip
- Oval marking spot
Ideal beaker for general laboratory procedures.
Customers Who Bought This Also Bought