Order Entry
ContactUsLinkComponent
DISH, TPX, STAINING
SP Bel-Art® Slide Staining and Storage System, Bel-Art Products, a part of SP
# de catalogue 13197-274
Fournisseur:  Avantor
DISH, TPX, STAINING
SP Bel-Art® Slide Staining and Storage System, Bel-Art Products, a part of SP
# de catalogue 13197-274
Fournisseur:  Avantor

Spécifications

  • Description:
    Staining Dish
  • Dimensions:
    9.4L x 8.1W x 4.9H cm (3¹¹⁄₁₆ x 3³⁄₁₆ x 2")
  • Supplier No.:
    F441820000

Spécifications

A propos de cet article

Universal slide rack and staining dish for histology, microbiology, and other slide staining needs.

  • Tight lid helps minimize spills and evaporation
  • Dish is easy to clean
  • Cover will not jam in freezer storage
  • Lid inside has a built-in slide rest

Made of black polyoxymethylene (POM) plastic, which is easy-to-clean and resistant to chemicals used in staining procedures.

Staining Dish
Capacity: 250 mL; only 150 mL is needed when using slide rack; provides a savings on staining liquids, developing fluids, and intermediates
Tight lid helps minimize spills and evaporation
Lid inside has a built-in slide rest; also provides a compartment for a drying agent
Dish is easy-to-clean; cover will not jam in freezer storage

Slide Rack (shown inside Staining Dish)
Holds twenty-five, 25 x 75 mm (1 x 3") slides in numbered slots
Has open bottom for rapid draining and a handle so the rack can be lifted to add or remove slides without touching the staining solution
Maximum use temperature is 90°C (194°F)

Customers Who Bought This Also Bought