Specifications
- Description:Refrigerator/Freezer Plus™ Thermometer
- Temperature resolution:0.1°
- Temperature accuracy:±0.5°C
- Cat. no.:12777-838
- Probe:Bottle probe (glycol)
- Range:...50.00...70.00 °C (...58.00...158.00 °F)
Specifications
About this item
Accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.
- Bottle insulates sensor from rapid temperature changes
- Solid-state probe eliminates mercury contamination in refrigerators
- Triple display design
Triple display simultaneously shows minimum, maximum and current temperatures.
Minimum/maximum monitors high/low readings overnight, on weekends, and for any time period - a significant advantage over current reading-only glass thermometers. Range is -58 to 158°F and -50 to 70°C with a resolution of 0.1° and an accuracy of ± 0.5°C. Alarm feature provides alert when temperature rises above or falls below a set point. Alarm is programmable in 1° increments. Ten-foot micro-cable permits refrigerator doors to close on it.
Probe is sealed in a miniature bottle filled with nontoxic glycol. Solution is GRAS generally recognized as safe by the US FDA.