Accessories for VWR® Vortex Mixers
# de catalogue 12620-872
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:Head with rubber cover, 76 mm (3")
- Environmentally Preferable:Waste Reducing
- Environmental certification:RoHS
- Cat. no.:12620-872
Spécifications
A propos de cet article
Customers Who Bought This Also Bought