VWR® Weighing Paper
# de catalogue 12578-121
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Size:7.6×7.6 cm (3×3")
Spécifications
A propos de cet article
Weighing paper is designed for analytical weighing with minimal influence on analytical results for weighing and transporting sample.
- Clean and dust-free
- Lightweight
- Nitrogen-free
- Smooth surface
A smooth surface is ideal for transferring weighed material. Clean, dust-free sheets. Paper is lightweight, yet strong enough to support analytical size samples.
Packaging: All sizes of weighing papers are packaged 500 sheets per box.
Customers Who Bought This Also Bought