ContactUsLinkComponent
VWR® Disposable Aluminum Weighing and Drying Pans
  12177-305
VWR* Disposable Aluminum Weighing Pan
VWR® Disposable Aluminum Weighing and Drying Pans
  12177-305
  • Capacity:
    40 ml
  • Description:
    Weighing Pans
  • Cat. no.:
    12177-305
  • I.D.×D:
    7×0.6 cm (2¾×¼")
These oil-free pans are ideal for general-purpose weighing, dispensing, storing, and drying applications.
  • Sturdy construction
  • Compatible with most balances and dryers
  • Ideal for general-purpose applications
  • Disposable
  • Aluminum construction
Sturdy, with a slight taper for easy stacking. The 7 cm (23/4") I.D. pan is ideal for enclosed balances and weighing equipment with a 7 to 9.5 cm (23/4 to 33/4") diameter head.
Customers Who Bought This Also Bought