Order Entry
ContactUsLinkComponent
Foam Rubber Squeegee Blade
Quick-Dry Foam Squeegees, Perfex
# de catalogue 11311-120
Fournisseur:  Perfex
Small Business Enterprise
Foam Rubber Squeegee Blade
Quick-Dry Foam Squeegees, Perfex
# de catalogue 11311-120
Fournisseur:  Perfex
Small Business Enterprise

Spécifications

  • Description:
    Foam Squeegee Blade
  • Cat. no.:
    11311-120
  • Length:
    45.7 cm (18")
  • Supplier No.:
    9718

Spécifications

A propos de cet article

Twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces

Note: Not recommended for abrasive surfaces.

Feature a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, nonmarring, and easy to clean with warm water. Resistant to water, oil, grease, detergents, sanitizers, and solvents.

Ordering information: Polymer adjustable handle sold separately.
Customers Who Bought This Also Bought