SP Bel-Art Odo-Clave® Autoclave Deodorant Pads, Bel-Art Products, a part of SP
# de catalogue 11215-983
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Cat. no.:11215-983
- Scent:Cherry
- Supplier No.:131980001
Spécifications
A propos de cet article
Heat-released deodorant pads neutralize autoclave odors with one of four pleasant scents.
- Effectively neutralize autoclave odors
- Choice of pleasant scents
- Long lifespan, especially if stored properly
- Easy to apply prior to autoclaving
Simply open foil pouch and place pad in bag prior to autoclaving
Also available in triple strength clove that releases three times the deodorant of the standard pads while costing less than two standard pads.
Pads have a three-year lifespan but will last longer if stored in a sealed container.
Customers Who Bought This Also Bought